Learn More
Medchemexpress LLC Sulfo-SANPAH | 102568-43-4 | MFCD00054986 | 99.8% | 470.41 g/mol | C16H18N6O9S | 5 MG
SDP
Catalog No. 5000221921
Encompass_Preferred
Sulfo-SANPAH is a heterobifunctional, photo-reactive cross-linker that couples primary amines to surfaces using an N-hydroxysuccinimide (NHS) ester and a UV-activated nitrophenyl azide. It is water-soluble and widely used to immobilize extracellular matrix proteins onto hydrogels and other substrates for cell culture and biomaterials applications.
  • Heterobifunctional reagent enabling covalent coupling via an NHS ester and a photoactivated azide.
  • Water-soluble sulfonate improves solubility for aqueous surface modification.
  • Photoactivation allows spatially controlled immobilization using UV light.
  • High purity supports reproducible surface functionalization.
  • Suitable for immobilizing extracellular matrix proteins onto hydrogels and biomaterials.
Catalog No. 50-002-21921 Supplier Medchemexpress LLC Supplier No. HY1373835MG
Sulfo-SANPAH is a heterobifunctional, photo-reactive cross-linker that couples primary amines to surfaces using an N-hydroxysuccinimide (NHS) ester and a UV-activated nitrophenyl azide. It is water-soluble and widely used to immobilize extracellular matrix proteins onto hydrogels and other substrates for cell culture and biomaterials applications.
  • Heterobifunctional reagent enabling covalent coupling via an NHS ester and a photoactivated azide.
  • Water-soluble sulfonate improves solubility for aqueous surface modification.
  • Photoactivation allows spatially controlled immobilization using UV light.
  • High purity supports reproducible surface functionalization.
  • Suitable for immobilizing extracellular matrix proteins onto hydrogels and biomaterials.