Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sulfo-Sanpah (Sulfosuccinimidyl 6-(4'-Azido-2'-Nitrophenylamino)Hexanoate)

Catalog No. 80333250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-333-250MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 80-333-250MG Supplier Sigma Aldrich Supplier No. 80333250MG
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

ANB-NOS and Sulfo-SANPAH are heterobifunctional crosslinkers that contain an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide. NHS esters react efficiently with primary amino groups (-NH2) in pH 7-9 buffers to form stable amide bonds. When exposed to UV light nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines).

Specifications

CAS 1329422-26-5
Melting Point 85°C to 90°C
Molecular Formula C19H22NO3PS
Quantity 100 mg
Synonym Exo-4-Methoxyphenyl Kwon [2.2.1] Bicyclic Phosphine; (1S,4S,5R)-5-(4-methoxyphenyl)-2-tosyl-2-aza-5-phosphabicyclo[2.2.1]heptane
Molecular Weight (g/mol) 375.42 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.