Learn More
CUBE BIOTECH Nanodisc Copolymer Sulfo-SMALP
Supplier:  CUBE BIOTECH 18523
Encompass
10x1g of nanodisc copolymer Sulfo-SMALO or the stabilization of membrane proteins outside the cell
Catalog No. NC2684573