Learn More
Aat Bioquest Sulfo-SMCC [4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester, sodium salt] *CAS#: 92921-24-9*
SDP
Catalog No. 501953237
Encompass_Preferred
Sulfo-SMCC is the water-soluble version of SMCC. It contains an amine-reactive sulfo-succinimidyl ester (SSE) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds.
Catalog No. 50-195-3237 Supplier Aat Bioquest Supplier No. 4504
Sulfo-SMCC is the water-soluble version of SMCC. It contains an amine-reactive sulfo-succinimidyl ester (SSE) and a sulfhydryl-reactive maleimide group. NHS esters react with primary amines at pH 7-9 to form stable amide bonds.