Learn More
Abcam Sulfo-SMCC, amine-to-sulfhydryl crosslinking agent, 50MG
Catalog No. 502204954
Encompass_Preferred
MW 436.4 Da, Purity >90%. Amine-to-sulfhydryl crosslinking agent. Contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length (8.3 angstroms) cyclohexane spacer arm. Introduces no more than two maleimide groups thus keeping protein modification low and preserving function.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4954 Supplier Abcam Supplier No. AB14560750MG
MW 436.4 Da, Purity >90%. Amine-to-sulfhydryl crosslinking agent. Contains NHS-ester and maleimide reactive groups at opposite ends of a medium-length (8.3 angstroms) cyclohexane spacer arm. Introduces no more than two maleimide groups thus keeping protein modification low and preserving function.
The product is subject to the following: Abcam Restricted Use Statement