Learn More
Medchemexpress LLC HY-D0975 10mg Medchemexpress, Sulfo-SMCC (sodium) CAS:92921-24-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD097510MG
Encompass_Preferred
Medchemexpress, HY-D0975 10mg Sulfo-SMCC (sodium) CAS:92921-24-9 Sulfo-SMCC sodium is a commonly used hetero-bifunctional crosslinker bearing N-hydroxysuccinimide (NHS) ester and maleimide groups to react with primary amines and sulfhydryl groups, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3222