Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfo SMCC R-phycoerythrin | 10 MG
SDP

Catalog No. 5000297566
Encompass_Preferred

Sulfo SMCC R-phycoerythrin is a liquid conjugate of the Sulfo-SMCC crosslinker and R-phycoerythrin supplied for labeling proteins with red/orange fluorescence in research applications.

  • Form: liquid solution.
  • Pack size: 10 mg (20 mg/mL · 500 μL).
  • Storage: store at -20 °C and protect from light.
  • Applications: protein labeling for fluorescence detection and flow cytometry.
  • Intended use: research use only.

Catalog No. 50-002-97566 Supplier Medchemexpress LLC Supplier No. HYD239610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfo SMCC R-phycoerythrin is a liquid conjugate of the Sulfo-SMCC crosslinker and R-phycoerythrin supplied for labeling proteins with red/orange fluorescence in research applications.

  • Form: liquid solution.
  • Pack size: 10 mg (20 mg/mL · 500 μL).
  • Storage: store at -20 °C and protect from light.
  • Applications: protein labeling for fluorescence detection and flow cytometry.
  • Intended use: research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.