Learn More
TARGETMOL CHEMICALS INC SULFO-SMCC SODIUM 200MG
SDP
Catalog No. 502581069
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulfo-SMCC sodium is a commonly used hetero-bifunctional non-cleavable ADC crosslinker bearing N-hydroxysuccinimide (NHS) ester and maleimide groups to react with primary amines and sulfhydryl groups respectively. purity: 99%
Catalog No. 50-258-1069 Supplier TARGETMOL CHEMICALS INC Supplier No. T18728200MG
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulfo-SMCC sodium is a commonly used hetero-bifunctional non-cleavable ADC crosslinker bearing N-hydroxysuccinimide (NHS) ester and maleimide groups to react with primary amines and sulfhydryl groups respectively. purity: 99%