Learn More
Medchemexpress LLC Sulfo-SMCC (sodium) | 92921-24-9 | 99.86% | 5 MG
SDP
Catalog No. 5000410605
Encompass_Preferred
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker. It contains N-hydroxysuccinimide (NHS) ester and maleimide groups which react with primary amines and sulfhydryl groups, respectively.
  • Hetero-bifunctional, noncleavable ADC crosslinker.
  • Contains N-hydroxysuccinimide (NHS) ester and maleimide groups.
  • Reacts with primary amines and sulfhydryl groups.
  • Inhibits the end-to-end annealing of stabilized microtubules.
  • Microtubules treated with Sulfo-SMCC show a constant mean length, independent of the incubation time.
Catalog No. 50-004-10605 Supplier Medchemexpress LLC Supplier No. HYD09755MG
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker. It contains N-hydroxysuccinimide (NHS) ester and maleimide groups which react with primary amines and sulfhydryl groups, respectively.
  • Hetero-bifunctional, noncleavable ADC crosslinker.
  • Contains N-hydroxysuccinimide (NHS) ester and maleimide groups.
  • Reacts with primary amines and sulfhydryl groups.
  • Inhibits the end-to-end annealing of stabilized microtubules.
  • Microtubules treated with Sulfo-SMCC show a constant mean length, independent of the incubation time.