Learn More
Medchemexpress LLC Sulfo-SMCC sodium | 92921-24-9 | 99.9% | 25 MG
SDP
Catalog No. 5000412060
Encompass_Preferred
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker. It features N-hydroxysuccinimide (NHS) ester and maleimide groups, which react with primary amines and sulfhydryl groups, respectively. This crosslinker has been shown to inhibit the end-to-end annealing of stabilized microtubules.
  • Hetero-bifunctional, noncleavable ADC crosslinker
  • Contains N-hydroxysuccinimide (NHS) ester and maleimide groups
  • Reacts with primary amines and sulfhydryl groups
Catalog No. 50-004-12060 Supplier Medchemexpress LLC Supplier No. 25MGHYD0975
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker. It features N-hydroxysuccinimide (NHS) ester and maleimide groups, which react with primary amines and sulfhydryl groups, respectively. This crosslinker has been shown to inhibit the end-to-end annealing of stabilized microtubules.
  • Hetero-bifunctional, noncleavable ADC crosslinker
  • Contains N-hydroxysuccinimide (NHS) ester and maleimide groups
  • Reacts with primary amines and sulfhydryl groups