Learn More
Medchemexpress LLC Sulfo-SMCC sodium | 92921-24-9 | 99.9% | 500 MG
SDP
Catalog No. 5000409953
Encompass_Preferred
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker. It contains N-hydroxysuccinimide (NHS) ester and maleimide groups, which allow it to react with primary amines and sulfhydryl groups, respectively. This product is intended for research use only and is not sold to patients.
  • Hetero-bifunctional
  • Noncleavable ADC crosslinker
  • Bears N-hydroxysuccinimide (NHS) ester group
  • Bears maleimide group
  • Reacts with primary amines and sulfhydryl groups
Catalog No. 50-004-09953 Supplier Medchemexpress LLC Supplier No. HYD0975500MG
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker. It contains N-hydroxysuccinimide (NHS) ester and maleimide groups, which allow it to react with primary amines and sulfhydryl groups, respectively. This product is intended for research use only and is not sold to patients.
  • Hetero-bifunctional
  • Noncleavable ADC crosslinker
  • Bears N-hydroxysuccinimide (NHS) ester group
  • Bears maleimide group
  • Reacts with primary amines and sulfhydryl groups