Learn More
Cayman Chemical Sulfo-SMCC (sodium salt), Purity: ≥95%, CAS: 92921-24-9, 100 mg
SDP
Catalog No. 502911386
Encompass_Preferred
A water-soluble protein cross-linking reagent; has been used to conjugate anti-PD-L1 antibodies to the surface of platelets and Golgi-targeting peptides to platelet microparticle-mimetic nanoplatforms
Catalog No. 50-291-1386 Supplier Cayman Chemical Supplier No. 35692100 MG
A water-soluble protein cross-linking reagent; has been used to conjugate anti-PD-L1 antibodies to the surface of platelets and Golgi-targeting peptides to platelet microparticle-mimetic nanoplatforms