Learn More
Bioworld Sulfo-SMCC, 100 mg
Supplier:  Bioworld 218100271
Encompass_Preferred
Sulfo-SMCC (Sulfosuccinimidyl-4-N-maleimidomethyl) cyclohexane-1-carboxylate)Synonym: 4-9N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodiumPrimary amine and sulfydryl reactiveEnzyme-antibody conjugationStable maleimide group References: 1) Hushida, S., et al. (1984) J. Applied Biochem. 56, 56-63.2) Staros, J.V. and Anjanejulu, P.S. (1989) Methods Enzymol. 172, 609.
Catalog No. 50-199-274