Learn More
Bioworld Sulfo-SMPB, 100 mg
Supplier:  Bioworld 218100281
Encompass_Preferred
Sulfo SMPB, Sulfosuccinimidyl 4-(p-maleimidophenyl) ButyratePrimary amine and sulfhydryl reactiveSpacer arm lenght: 11.6A°Water solubleReferences: 1.) Bangs, J.D. et al. (1986) J. Cell Biol. 103, 255-263. 2.) Myers, D.E., et al. (1989) J. Immunol. Methods 121, 129-142.M.W. = 458.4
Catalog No. 50-199-275