Learn More
Medchemexpress LLC Sulfo-SPDB | 1193111-39-5 | MFCD20227457 | 92.1% | 406.45 g·mol⁻¹ | C13H14N2O7S3 | 10 MG
SDP
Catalog No. 5000211202
Encompass_Preferred
sulfo-SPDB is a cleavable, disulfide-based ADC linker for research use, designed to connect cytotoxic payloads to antibodies and enable intracellular release. The reagent includes a sulfonate group for improved aqueous solubility and a pyridyl disulfide moiety that supports reductive cleavage under intracellular conditions.
  • Enables controlled release of payloads via reductive cleavage.
  • Improved aqueous solubility due to sulfonate functionality.
  • Suitable for use in antibody-drug conjugate synthesis workflows.
  • Compatible with common bioconjugation and purification methods.
  • Provided with product documentation and lot-specific purity information.
Catalog No. 50-002-11202 Supplier Medchemexpress LLC Supplier No. HY10115110MG
sulfo-SPDB is a cleavable, disulfide-based ADC linker for research use, designed to connect cytotoxic payloads to antibodies and enable intracellular release. The reagent includes a sulfonate group for improved aqueous solubility and a pyridyl disulfide moiety that supports reductive cleavage under intracellular conditions.
  • Enables controlled release of payloads via reductive cleavage.
  • Improved aqueous solubility due to sulfonate functionality.
  • Suitable for use in antibody-drug conjugate synthesis workflows.
  • Compatible with common bioconjugation and purification methods.
  • Provided with product documentation and lot-specific purity information.