Learn More
Medchemexpress LLC HY-101151 5mg , sulfo-SPDB CAS:1193111-39-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1011515MG
Encompass_Preferred
HY-101151 5mg sulfo-SPDB CAS: 1193111-39-5 sulfo-SPDB is a cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1615