Learn More
Medchemexpress LLC Sulfo-SPP | 452072-27-4 | 97.1% | 420.48 g·mol⁻1 | C14H16N2O7S3 | 5 MG
SDP
Catalog No. 5000203956
Encompass_Preferred
Sulfo-SPP is a heterobifunctional, thiol-cleavable, membrane-impermeable crosslinker used as an ADC linker in bioconjugation research. It reacts selectively with primary amines and contains a cleavable disulfide linkage that enables release under reducing conditions. Supplied for research use only.
  • Heterobifunctional thiol-cleavable crosslinker.
  • Membrane impermeable sulfonate group improves aqueous solubility.
  • Cleavable disulfide linkage for controlled release under reducing conditions.
  • Suitable for antibody-drug conjugate and bioconjugation applications.
  • Available in small pack sizes for research use.
Catalog No. 50-002-03956 Supplier Medchemexpress LLC Supplier No. HY1293775MG
Sulfo-SPP is a heterobifunctional, thiol-cleavable, membrane-impermeable crosslinker used as an ADC linker in bioconjugation research. It reacts selectively with primary amines and contains a cleavable disulfide linkage that enables release under reducing conditions. Supplied for research use only.
  • Heterobifunctional thiol-cleavable crosslinker.
  • Membrane impermeable sulfonate group improves aqueous solubility.
  • Cleavable disulfide linkage for controlled release under reducing conditions.
  • Suitable for antibody-drug conjugate and bioconjugation applications.
  • Available in small pack sizes for research use.