Learn More
Abcam Sulfobetaine 10, Zwitterionic detergent, 1G
Catalog No. 502704684
Encompass_Preferred
MW 307.48 g/mol, Purity >98%. Detergent used in the solubilization of oleosins. Synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range. This property is attributed to the presence of a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding to either anionic or cationic compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4684 Supplier Abcam Supplier No. AB1463631G
MW 307.48 g/mol, Purity >98%. Detergent used in the solubilization of oleosins. Synthetic zwitterionic detergent, which unlike other amphoteric surfactants, retains its zwitterionic character over a broad pH range. This property is attributed to the presence of a strongly basic quaternary ammonium ion and an acidic sulfonate ion of equal strength. This ionic balance presumably prevents irreversible binding to either anionic or cationic compounds.
The product is subject to the following: Abcam Restricted Use Statement