Learn More
Medchemexpress LLC Sulfobetaine-14 Sta 250mg
SDP
Catalog No. 5000205786
Encompass_Preferred
Sulfobetaine-14 (Standard) is the analytical standard of Sulfobetaine-14 This product is intended for research and analytical applications Zwittergent 3-14 (DMAPS) is a zwitterionic detergent commonly used in biochemistry and molecular biology for the solubilization and purification of membrane-bound proteins and other hydrophobic biomolecules which have both hydrophilic and hydrophobic moieties so that it has good detergency properties making it suitable for stabilizing membrane proteins in aqueous solutions In addition DMAPS has been used in various techniques such as electrophoresis and chromatography for the separation and analysis of biomolecules The long The hydrocarbon chains provide it with good membrane penetration and solubilization capabilities while the sulfonate and quaternary ammonium groups ensure water solubility and charge neutrality
Catalog No. 50-002-05786 Supplier Medchemexpress LLC Supplier No. 250MGHYW099581R
Sulfobetaine-14 (Standard) is the analytical standard of Sulfobetaine-14 This product is intended for research and analytical applications Zwittergent 3-14 (DMAPS) is a zwitterionic detergent commonly used in biochemistry and molecular biology for the solubilization and purification of membrane-bound proteins and other hydrophobic biomolecules which have both hydrophilic and hydrophobic moieties so that it has good detergency properties making it suitable for stabilizing membrane proteins in aqueous solutions In addition DMAPS has been used in various techniques such as electrophoresis and chromatography for the separation and analysis of biomolecules The long The hydrocarbon chains provide it with good membrane penetration and solubilization capabilities while the sulfonate and quaternary ammonium groups ensure water solubility and charge neutrality