Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfobromophthalein disodium salt | 71-67-0 | 99.8% | 500 MG
SDP

Catalog No. 5000410502
Encompass_Preferred

Sulfobromophthalein (Bromosulfophthalein) disodium salt is an organic anion dye used in the study of a variety of membrane carriers expressed in animal tissues and involved in transport of agents and metabolites. It is a synthetic dye introduced in 1925 for assessing liver function and also acts as a pH indicator dye. It has been commonly used as both a substrate and inhibitor of organic anionic transporting polypeptide 1B1 (OATP1B1), OATP1B3, OATP1A2, OATP2B1, and MDR2. For research use only.

  • Used for liver function assessment
  • Acts as a pH indicator dye
  • Substrate and inhibitor for OATP1B1, OATP1B3, OATP1A2, OATP2B1, and MDR2
  • Purity: 99.77%
  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 838.00
  • Formula: C20H8Br4Na2O10S2

Catalog No. 50-004-10502 Supplier Medchemexpress LLC Supplier No. HYD0217500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfobromophthalein (Bromosulfophthalein) disodium salt is an organic anion dye used in the study of a variety of membrane carriers expressed in animal tissues and involved in transport of agents and metabolites. It is a synthetic dye introduced in 1925 for assessing liver function and also acts as a pH indicator dye. It has been commonly used as both a substrate and inhibitor of organic anionic transporting polypeptide 1B1 (OATP1B1), OATP1B3, OATP1A2, OATP2B1, and MDR2. For research use only.

  • Used for liver function assessment
  • Acts as a pH indicator dye
  • Substrate and inhibitor for OATP1B1, OATP1B3, OATP1A2, OATP2B1, and MDR2
  • Purity: 99.77%
  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 838.00
  • Formula: C20H8Br4Na2O10S2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.