Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfobromophthalein Disodium Salt Hydrate, Sigma-Aldrich

Catalog No. 01258914 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-914 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-914 Supplier MilliporeSigma Supelco Supplier No. S02525G
Add to Cart
Add to Cart

Sulfobromophthalein disodium salt hydrate has been used as a positive control to study the interaction of 11-keto-B-boswellic acid (KBA) and 3-acetyl-11-keto-B-boswellic acid (AKBA) with organic anion transporter OATP1B3.

TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
Form Powder
Quantity 5 g
Substrate Properties Chemical Substrate
Product Type Substrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.