Learn More
Medchemexpress LLC Sulfometuron-methyl | 74222-97-2 | 97.4% | 25 MG
SDP
Catalog No. 5000413846
Encompass_Preferred
Sulfometuron-methyl is a herbicide and a powerful inhibitor of the enzyme acetolactate synthase (ALS). It demonstrates both antibacterial and antifungal properties.
  • Inhibits acetolactate synthase (ALS).
  • Exhibits antibacterial activity.
  • Exhibits antifungal activity.
  • Capable of inhibiting the activity of ALS II in Salmonella typhimurium.
  • Capable of inhibiting the activity of ALS III in Escherichia coli.
Catalog No. 50-004-13846 Supplier Medchemexpress LLC Supplier No. 25MGHYB1852
Sulfometuron-methyl is a herbicide and a powerful inhibitor of the enzyme acetolactate synthase (ALS). It demonstrates both antibacterial and antifungal properties.
  • Inhibits acetolactate synthase (ALS).
  • Exhibits antibacterial activity.
  • Exhibits antifungal activity.
  • Capable of inhibiting the activity of ALS II in Salmonella typhimurium.
  • Capable of inhibiting the activity of ALS III in Escherichia coli.