Learn More
Cayman Chemical Sulfonadyn-47, Purity: ≥98%, 10 mg
SDP
Catalog No. 502917185
Encompass_Preferred
A dynamin inhibitor; inhibits dynamin GTPase activity in a cell-free assay (IC50 = 3.5 µM); inhibits clathrin-mediated endocytosis in U2OS cells (IC50 = 27.3 µM), as well depolarization-stimulated uptake of FM4-64 in isolated rat brain synaptosomes (IC50 = 12.3 µM); has anticonvulsant activity in the 6 Hz corneal kindling mouse model of seizures at 30 and 60 mg/kg
Catalog No. 50-291-7185 Supplier Cayman Chemical Supplier No. 3940210 MG
A dynamin inhibitor; inhibits dynamin GTPase activity in a cell-free assay (IC50 = 3.5 µM); inhibits clathrin-mediated endocytosis in U2OS cells (IC50 = 27.3 µM), as well depolarization-stimulated uptake of FM4-64 in isolated rat brain synaptosomes (IC50 = 12.3 µM); has anticonvulsant activity in the 6 Hz corneal kindling mouse model of seizures at 30 and 60 mg/kg