Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chiron Sulfonamide, N-methyl, N-hydroxyethyl Standards
SDP

Catalog No. 148461150ME Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-846-1150ME 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-846-1150ME Supplier Chiron Supplier No. 14846.1150ME

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material N-(2-Hydroxyethyl)-N-methylperfluorooctanesulfonamide-d7
CAS 1265205-95-5
Concentration 50 μg/mL
Synonym N-MeFOSE-d7
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.