Learn More
Description
- Various singles available
- Solutions in assorted solvents
- Useful for various matrices
- Custom standards of other PFAS compounds available on demand
Specifications
Specifications
| Chemical Name or Material | N-Methyl-n-perfluorobutanesulfonamide-d3 |
| Concentration | 50 μg/mL |
| Synonym | MeFBSA-d3 |
| Type | Reference Material |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
