Learn More
American Research Products Inc Sulfonamides SAs residue ELI
Catalog No. 5000329238
Encompass
Sulfonamides (SAs) residue ELISA kit from CUSABIO Detection Range 2 ppb- 162 ppb Sensitivity 2 ppb Sample Tissue(Chicken pork liver) pig urine milk etc Method
Catalog No. 50-003-29238 Supplier American Research Products Inc Supplier No. CSBE12094F
Sulfonamides (SAs) residue ELISA kit from CUSABIO Detection Range 2 ppb- 162 ppb Sensitivity 2 ppb Sample Tissue(Chicken pork liver) pig urine milk etc Method