Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfone, bis((4-chloroacetylamino)phenyl). | 17328-16-4 | MFCD00028183 | 98.0% | 401.26 g/mol | C16H14Cl2N2O4S | 1 ML
SDP

Catalog No. 5000240406
Encompass_Preferred

TC-E 5003 is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor supplied as a ready-to-use 10 mM solution in DMSO for research use in biochemical and cell-based assays. Handle and store according to laboratory safety and storage guidelines.

  • Selective PRMT1 inhibitor with reported IC50 ≈ 1.5 μM.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Purity 98.0% by HPLC.
  • Molecular weight 401.26 g/mol.
  • Store solution at -80°C for long-term stability; short-term storage at -20°C.

Catalog No. 50-002-40406 Supplier Medchemexpress LLC Supplier No. HY10757410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TC-E 5003 is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor supplied as a ready-to-use 10 mM solution in DMSO for research use in biochemical and cell-based assays. Handle and store according to laboratory safety and storage guidelines.

  • Selective PRMT1 inhibitor with reported IC50 ≈ 1.5 μM.
  • Supplied as a 10 mM solution in DMSO, 1 mL vial.
  • Purity 98.0% by HPLC.
  • Molecular weight 401.26 g/mol.
  • Store solution at -80°C for long-term stability; short-term storage at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.