Learn More
Medchemexpress LLC HY-139361 10mg , Sulfopin CAS:2451481-08-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13936110MG
Encompass_Preferred
HY-139361 10mg Sulfopin CAS: 2451481-08-4 Sulfopin (PIN1-3) is a highly selective covalent inhibitor of Pin1 with an apparent Ki of 17 nM. Sulfopin blocks Myc-driven tumors in vivo. The peptidyl-prolyl isomerase, Pin1, is exploited in cancer to activate oncogenes and inactivate tumor suppressors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0827