Learn More
Apexbio Technology LLC Sulforaphane(Synonyms: Broccoli sulforaphane, DL-Sulforaphane, R-Sulforaphane, 1-Isothiocyanato-4-(methylsulfinyl)butane), 10mg, CAS: 4478-93-7.
SDP
Supplier:  Apexbio Technology LLC C473310
Encompass_Preferred
Sulforaphane (CAS 4478-93-7) is a naturally occurring isothiocyanate abundantly found in vegetables especially broccoli It acts primarily through activation of the Keap1-Nrf2 signaling pathway enhancing cellular defense against oxidative and electrophilic stress Sulforaphane has demonstrated antiproliferative activity in vitro as shown by dose-dependent cell cycle arrest at G2/M and apoptosis induction in HT29 human colon carcinoma cells Such effects involve elevated cyclin A and B1 levels increased pro-apoptotic Bax expression mitochondrial cytochrome c release and poly(ADP-ribose) polymerase cleavage These properties make sulforaphane relevant for investigating cancer chemoprevention and cellular stress responses
Catalog No. 50-000-61183