Learn More
Selleck Chemical LLC Sulforaphane
SDP
Catalog No. 5000003293
Encompass_Preferred
Sulforaphane is a naturally occurring isothiocyanate derived from the consumption of cruciferous vegetables such as broccoli cabbage and kale It is an inducer of Nrf2 Sulforaphane is also an inhibitor of histone deacetylase (HDAC) and NF- B Sulforaphane increases heme oxygenase-1 (HO-1) and reduces the levels of reactive oxygen species (ROS) Sulforaphane induces cell cycle arrest and apoptosis
Catalog No. 50-000-03293 Supplier Selleck Chemical LLC Supplier No. S577125MG
Sulforaphane is a naturally occurring isothiocyanate derived from the consumption of cruciferous vegetables such as broccoli cabbage and kale It is an inducer of Nrf2 Sulforaphane is also an inhibitor of histone deacetylase (HDAC) and NF- B Sulforaphane increases heme oxygenase-1 (HO-1) and reduces the levels of reactive oxygen species (ROS) Sulforaphane induces cell cycle arrest and apoptosis