Learn More
TARGETMOL CHEMICALS INC Sulforaphane 25MG
SDP
Catalog No. 502877298
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Sulforaphane is a natural isothiocyanate that activates Nrf2 and inhibits AMPK-dependent signaling. Sulforaphane has antitumor and anti-inflammatory activities by increasing the transcription of tumor suppressor proteins and inhibiting histone deacetylase activity. Purity 99.79%
Catalog No. 50-287-7298 Supplier TARGETMOL CHEMICALS INC Supplier No. TQ020725MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Sulforaphane is a natural isothiocyanate that activates Nrf2 and inhibits AMPK-dependent signaling. Sulforaphane has antitumor and anti-inflammatory activities by increasing the transcription of tumor suppressor proteins and inhibiting histone deacetylase activity. Purity 99.79%