Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulforaphane | 4478-93-7 | MFCD00198068 | 98.9% | 177.29 g/mol | C6H11NOS2 | 5 MG
SDP

Catalog No. 5000222922
Encompass_Preferred

Sulforaphane is a research-grade isothiocyanate that activates the Keap1/Nrf2/ARE antioxidant pathway and inhibits histone deacetylases. Supplied as a high-purity powder, it is intended for use in in vitro and in vivo studies of antioxidant response, anticancer activity, and anti-inflammatory mechanisms.

  • High purity: 98.9%.
  • CAS number: 4478-93-7.
  • Molecular formula: C6H11NOS2; MW: 177.29 g/mol.
  • Suitable for in vitro and in vivo applications.
  • Storage: powder -20°C; in solvent -80°C for long-term storage.
  • Available in small research quantities, including 5 mg.

Catalog No. 50-002-22922 Supplier Medchemexpress LLC Supplier No. HY137555MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulforaphane is a research-grade isothiocyanate that activates the Keap1/Nrf2/ARE antioxidant pathway and inhibits histone deacetylases. Supplied as a high-purity powder, it is intended for use in in vitro and in vivo studies of antioxidant response, anticancer activity, and anti-inflammatory mechanisms.

  • High purity: 98.9%.
  • CAS number: 4478-93-7.
  • Molecular formula: C6H11NOS2; MW: 177.29 g/mol.
  • Suitable for in vitro and in vivo applications.
  • Storage: powder -20°C; in solvent -80°C for long-term storage.
  • Available in small research quantities, including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.