Learn More
Cayman Chemical Sulforaphane, 4478-93-7, 5 mg
SDP
Supplier:  Cayman Chemical 104965
Encompass_Preferred
Molecular Formula C6H11NOS2, Purity ≥98%, Formula Weight 177.3, Synonyms: SFN; An isothiocyanate that potently induces chemopreventative enzymes via Keap1-Nrf2 signaling and ARE-driven gene expression; at 15 M, inhibits class I and II HDAC activity and suppresses tumor growth selectively in cancerous prostate epithelial cells without affecting normal cells.
Catalog No. NC1387095