Learn More
Cayman Chemical Sulforaphane Inhibitors
SDP
Catalog No. 502888942
Encompass_Preferred
An isothiocyanate that potently induces chemopreventative enzymes via Keap1-Nrf2 signaling and ARE-driven gene expression; at 15 μM, inhibits class I and II HDAC activity and suppresses tumor growth selectively in cancerous prostate epithelial cells without affecting normal cells
Catalog No. 50-288-8942 Supplier Cayman Chemical Supplier No. 1049650 MG
An isothiocyanate that potently induces chemopreventative enzymes via Keap1-Nrf2 signaling and ARE-driven gene expression; at 15 μM, inhibits class I and II HDAC activity and suppresses tumor growth selectively in cancerous prostate epithelial cells without affecting normal cells