Learn More
TARGETMOL CHEMICALS INC SULFORAPHENE 10MG
SDP
Catalog No. 502576179
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulforaphene is a natural product isolated from radish seeds exhibits an ED50 against velvetleaf seedlings approximately 2 x 10 -4 M. Sulforaphene promotes cancer cells apoptosis and inhibits migration via inhibiting EGFR p-ERK1/2 NF‐(kappa)B and other signals. purity: 99%
Catalog No. 50-257-6179 Supplier TARGETMOL CHEMICALS INC Supplier No. TL001610MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulforaphene is a natural product isolated from radish seeds exhibits an ED50 against velvetleaf seedlings approximately 2 x 10 -4 M. Sulforaphene promotes cancer cells apoptosis and inhibits migration via inhibiting EGFR p-ERK1/2 NF‐(kappa)B and other signals. purity: 99%