Learn More
Cayman Chemical Sulforhodamine B (sodium salt) (technical grade), Purity: ≥75%, CAS: 3520-42-1, 10 mg
SDP
Catalog No. 502990714
Encompass_Preferred
An aminoxanthene dye; binds basic amino acid residues under mild acidic conditions, which can be quantified by colorimetric detection at 510 nM
Catalog No. 50-299-0714 Supplier Cayman Chemical Supplier No. 3153910 MG
An aminoxanthene dye; binds basic amino acid residues under mild acidic conditions, which can be quantified by colorimetric detection at 510 nM