Learn More
AdipoGen Sulforhodamine 101,Chemical. CAS: 60311-02-6
SDP
Catalog No. 501147241
Encompass_Preferred
Sulforhodamine 101, AdipoGen Life Sciences. Chemical. CAS: 60311-02-6. Formula: C31H30N2O7S2. MW: 606.71. Synthetic Sulforhodamine 101 is a non-fixable red fluorescent dye used as a specific marker for astrocytes and an activity-dependent probe for monitoring regulated exocytosis. In addition to labeling astrocytes, it also labels myelinating oligodendrocytes. Has been reported to affect excitability of neurons and should therefore be used with caution. Key precursor used for preparing rhodamine 101 sulfonyl chloride (also called Texas Red). Spectral data: lambdaex 586 nm| lambdaem 605 nm in H2O and lambdaex 582-588 nm| lambdaem 601-609 nm in pH7 buffer.
Catalog No. 50-114-7241 Supplier AdipoGen Supplier No. CDXS0025M100
Sulforhodamine 101, AdipoGen Life Sciences. Chemical. CAS: 60311-02-6. Formula: C31H30N2O7S2. MW: 606.71. Synthetic Sulforhodamine 101 is a non-fixable red fluorescent dye used as a specific marker for astrocytes and an activity-dependent probe for monitoring regulated exocytosis. In addition to labeling astrocytes, it also labels myelinating oligodendrocytes. Has been reported to affect excitability of neurons and should therefore be used with caution. Key precursor used for preparing rhodamine 101 sulfonyl chloride (also called Texas Red). Spectral data: lambdaex 586 nm| lambdaem 605 nm in H2O and lambdaex 582-588 nm| lambdaem 601-609 nm in pH7 buffer.