Learn More
AdipoGen Sulforhodamine 101, 60311-02-6, MFCD00012407, 500 mg
SDP
Catalog No. 501147242
Encompass_Preferred
MF: C31H30N2O7S2, MW: 606.71, ≥99%, Appearance: Green crystalline. Sulforhodamine 101 is a non-fixable red fluorescent dye used as a specific marker for astrocytes and an activity-dependent probe for monitoring regulated exocytosis. In addition to labeling astrocytes, it also labels myelinating oligodendrocytes. Has been reported to affect excitability of neurons and should therefore be used with caution. Key precursor used for preparing rhodamine 101 sulfonyl chloride (also called Texas Red).
Catalog No. 50-114-7242 Supplier AdipoGen Supplier No. CDXS0025M500
MF: C31H30N2O7S2, MW: 606.71, ≥99%, Appearance: Green crystalline. Sulforhodamine 101 is a non-fixable red fluorescent dye used as a specific marker for astrocytes and an activity-dependent probe for monitoring regulated exocytosis. In addition to labeling astrocytes, it also labels myelinating oligodendrocytes. Has been reported to affect excitability of neurons and should therefore be used with caution. Key precursor used for preparing rhodamine 101 sulfonyl chloride (also called Texas Red).