Learn More
Biotium Sulforhodamine 101
Catalog No. 501964789
Encompass_Preferred
Sulforhodamine 101 has a longer absorption wavelength than does sulforhodamine B. Like sulforhodamine B and G, in addition to their potential use in cancer drug screening, these fluorescent dyes have been primarily used as polar tracers for the studies of neuronal cell morphology and cell-cell communication. Properties: Ex/Em = 586/605 nm; Red solid soluble in water; Store at room temperature; C31H30N2O7S2; MW: 607; [60311-02-6]
Catalog No. 50-196-4789 Supplier Biotium Supplier No. 80101
Sulforhodamine 101 has a longer absorption wavelength than does sulforhodamine B. Like sulforhodamine B and G, in addition to their potential use in cancer drug screening, these fluorescent dyes have been primarily used as polar tracers for the studies of neuronal cell morphology and cell-cell communication. Properties: Ex/Em = 586/605 nm; Red solid soluble in water; Store at room temperature; C31H30N2O7S2; MW: 607; [60311-02-6]