Learn More
Abcam Sulforhodamine 101, 5MG
Catalog No. 50996240
Encompass_Preferred
MW 606.7 Da, Purity >95%. Red fluorophore standard for neurological staining and enzyme assays. Used as polar tracers for the studies of neuronal cell morphology and cell-cell communications. Sulforhodamine 101 can label astrocytes and myelinating oligodendrocytes. It has a longer absorption wavelength than sulforhodamine B and G.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-240 Supplier Abcam Supplier No. AB2750865MG
MW 606.7 Da, Purity >95%. Red fluorophore standard for neurological staining and enzyme assays. Used as polar tracers for the studies of neuronal cell morphology and cell-cell communications. Sulforhodamine 101 can label astrocytes and myelinating oligodendrocytes. It has a longer absorption wavelength than sulforhodamine B and G.
The product is subject to the following: Abcam Restricted Use Statement