Learn More
Apexbio Technology LLC Sulforhodamine 101 60311-02-6 25mg
SDP
Catalog No. 5000091938
Encompass_Preferred
Sulforhodamine 101 (60311-02-6) is a red-emitting fluorescent dye primarily utilized as a selective marker for astrocytes Sulforhodamine 101 selectively accumulates in astrocytes enabling differentiation from surrounding neuronal populations In neurobiological studies using live tissue imaging including acute brain slice preparations the dye facilitates visualization of astroglial cell morphology and cellular dynamics Based on these properties Sulforhodamine 101 holds research potential in the study of astrocyte-specific responses morphological characteristics and cell cell interactions within neuronal tissues
Catalog No. 50-000-91938 Supplier Apexbio Technology LLC Supplier No. B776225
Sulforhodamine 101 (60311-02-6) is a red-emitting fluorescent dye primarily utilized as a selective marker for astrocytes Sulforhodamine 101 selectively accumulates in astrocytes enabling differentiation from surrounding neuronal populations In neurobiological studies using live tissue imaging including acute brain slice preparations the dye facilitates visualization of astroglial cell morphology and cellular dynamics Based on these properties Sulforhodamine 101 holds research potential in the study of astrocyte-specific responses morphological characteristics and cell cell interactions within neuronal tissues