Learn More
Biotium Sulforhodamine 101 cadaverine (Texas Red cadaverine)
Catalog No. 501964826
Encompass_Preferred
Sulforhodamine 101 cadaverine can be coupled to activated carboxy groups or sulfonyl chlorides. Properties: Ex/Em (MeOH) = 583/603 nm; Dark purple solid soluble in DMSO or DMF; Store at 4°C and protect from light; C 36 H 42 N 4 O 6 S 2; MW: 690.87
Catalog No. 50-196-4826 Supplier Biotium Supplier No. 90109
Sulforhodamine 101 cadaverine can be coupled to activated carboxy groups or sulfonyl chlorides. Properties: Ex/Em (MeOH) = 583/603 nm; Dark purple solid soluble in DMSO or DMF; Store at 4°C and protect from light; C 36 H 42 N 4 O 6 S 2; MW: 690.87