Learn More
Selleck Chemical LLC Sulforhodamine 101-E0366-5MG
SDP
Catalog No. 503137095
Encompass_Preferred
Sulforhodamine 101 (SR101) is a fluorescent dye and is widely used for astrocyte identification
Catalog No. 50-313-7095 Supplier Selleck Chemical LLC Supplier No. E03665MG
Sulforhodamine 101 (SR101) is a fluorescent dye and is widely used for astrocyte identification