Learn More
Biotium Sulforhodamine 101-PEO3-amine (Texas Red-PEO3-amine)
Catalog No. 501964827
Encompass_Preferred
Sulforhodamine 101-PEO 3 -amine has a long water-soluble spacer separating the dye and the amine group. The dye can be coupled to activated carboxy groups or sulfonyl chlorides.Properties: Ex/Em (MeOH) = 583/603 nm; Dark purple solid soluble in DMSO or DMF; Store at 4°C and protect from light; C 39 H 48 N 4 O 9 S 2; MW: 780.95
Catalog No. 50-196-4827 Supplier Biotium Supplier No. 90110
Add to Cart
Edge
Add to Cart
Sulforhodamine 101-PEO 3 -amine has a long water-soluble spacer separating the dye and the amine group. The dye can be coupled to activated carboxy groups or sulfonyl chlorides.Properties: Ex/Em (MeOH) = 583/603 nm; Dark purple solid soluble in DMSO or DMF; Store at 4°C and protect from light; C 39 H 48 N 4 O 9 S 2; MW: 780.95