Learn More
Aat Bioquest Sulforhodamine 101 sulfonyl chloride [Texas Red®] *CAS 82354-19-6*
SDP
Catalog No. 501952898
Encompass_Preferred
Sulforhodamine 101 acid chloride (also called Texas Red®) is the most popular labeling reagent of sulfonyl chloride. It is quite unstable in water, especially at the higher pH required for reaction with aliphatic amines.
Catalog No. 50-195-2898 Supplier Aat Bioquest Supplier No. 480
Sulforhodamine 101 acid chloride (also called Texas Red®) is the most popular labeling reagent of sulfonyl chloride. It is quite unstable in water, especially at the higher pH required for reaction with aliphatic amines.