Learn More
Aat Bioquest Sulforhodamine 101, sodium salt
SDP
Catalog No. 502979322
Encompass_Preferred
Sulforhodamine 101 is a fluorescence reference standard for measuring fluorescence quantum yield.
Catalog No. 50-297-9322 Supplier Aat Bioquest Supplier No. 71
Sulforhodamine 101 is a fluorescence reference standard for measuring fluorescence quantum yield.