Learn More
Apexbio Technology LLC Sulforhodamine B sodium salt 3520-42-1 1g
SDP
Catalog No. 5000093683
Encompass_Preferred
Sulforhodamine B sodium salt (CAS 3520-42-1) is a fluorescent dye used for quantification of total cellular protein Sulforhodamine B sodium salt functions by electrostatically binding to cellular protein residues In cell-based studies fluorescence intensity after washing off unbound dye reflects the protein content of cultured cells This principle is applied for protein quantification assays including cytotoxicity assessments and cell proliferation assays Based on these properties Sulforhodamine B sodium salt is utilized in cell biology and biomedical research applications
Catalog No. 50-000-93683 Supplier Apexbio Technology LLC Supplier No. A46191000
Sulforhodamine B sodium salt (CAS 3520-42-1) is a fluorescent dye used for quantification of total cellular protein Sulforhodamine B sodium salt functions by electrostatically binding to cellular protein residues In cell-based studies fluorescence intensity after washing off unbound dye reflects the protein content of cultured cells This principle is applied for protein quantification assays including cytotoxicity assessments and cell proliferation assays Based on these properties Sulforhodamine B sodium salt is utilized in cell biology and biomedical research applications