Learn More
Selleck Chemical LLC Sulforhodamine B sodium salt
SDP
Catalog No. 5000004065
Encompass_Preferred
Sulforhodamine B (SRB Acid Red 52 Kiton Red 620) sodium salt is a protein-specific fluorescent dye that is utilized to quantitate cell expansion growth and migration
Catalog No. 50-000-04065 Supplier Selleck Chemical LLC Supplier No. S5976200MG
Sulforhodamine B (SRB Acid Red 52 Kiton Red 620) sodium salt is a protein-specific fluorescent dye that is utilized to quantitate cell expansion growth and migration