Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfosalicylic Acid, 20% (w/v), Ricca Chemical
SDP

Catalog No. 813016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8130-16 500 mL Amber Glass Bottle
8130-32 1 L Amber Glass Bottle
8130-1 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 8130-16 Supplier Ricca Chemical Company Supplier No. 813016
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5

Specifications

Percent Purity 20%
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 500 mL
Chemical Name or Material Sulfosalicylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.