Learn More
Hach Company Sulfosalicylic Acid Powder Pillows, pk/100
Catalog No. 501995246 Shop All Hach Company Products
Encompass
Indicator for iron determination by EDTA titration
Catalog No. 50-199-5246 Supplier Hach Company Supplier No. 2081669
Indicator for iron determination by EDTA titration